KEY465195066 Key Organics / BIONET

1-(4-fluorobenzenesulfonyl)-N3,N3,N5,N5-tetramethyl-1H-1,2,4-triazole-3,5-diamine

  • MDL number MFCD00794193
  • Cas Number : 478032-15-4
  • Molecular Weight : 313.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/71982

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3M-728_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart