KEY465195067 Key Organics / BIONET

14,14-dimethyl-10-(4-nitrophenyl)-2,9-diazatricyclo[9.4.0.0^{3,8}]pentadeca-1(11),3,5,7-tetraen-12-one

  • MDL number MFCD00731281
  • Cas Number : 82408-04-6
  • Molecular Weight : 363.417

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/71983

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3M-729_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart