KEY465195069 Key Organics / BIONET

ethyl 4-(furan-2-yl)-2-methyl-5-oxo-1,4,5,6,7,8-hexahydroquinoline-3-carboxylate

  • MDL number MFCD00771032
  • Cas Number : 338415-01-3
  • Molecular Weight : 301.342

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/71987

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3M-734_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart