KEY465195070 Key Organics / BIONET

(2Z,3E)-2-(N-hydroxyimino)-N-phenyl-3-(2-phenylhydrazin-1-ylidene)butanamide

  • MDL number MFCD00794550
  • Molecular Weight : 296.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/71992

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3M-743_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart