KEY465195073 Key Organics / BIONET

4-[(1E)-{[(4-chlorophenyl)methoxy]imino}methyl]-2-methyl-6-oxo-1,4,5,6-tetrahydropyridine-3-carbonitrile

  • MDL number MFCD00975018
  • Cas Number : 338415-14-8
  • Molecular Weight : 303.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/72003

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3M-907_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart