KEY465195074 Key Organics / BIONET

(4E)-2-methyl-6-oxo-4-{[(3,4,5-trimethoxyphenyl)amino]methylidene}-1,4,5,6-tetrahydropyridine-3-carbonitrile

  • MDL number MFCD00975022
  • Cas Number : 338415-18-2
  • Molecular Weight : 329.356

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/72008

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3M-912_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart