KEY465195078 Key Organics / BIONET

[2-(2-chloro-6-fluorophenyl)cyclopropyl](4-fluorophenyl)methanone

  • MDL number MFCD01315517
  • Cas Number : 338415-54-6
  • Molecular Weight : 292.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/72048

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3N-003_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart