KEY465195079 Key Organics / BIONET

[(E)-[1-(pyridin-2-yl)ethylidene]amino]urea

  • MDL number MFCD00832644
  • Cas Number : 120446-04-0
  • Molecular Weight : 178.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/72050

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3N-005_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart