KEY465195082 Key Organics / BIONET

(2E)-3-(4-methoxyphenyl)-1-(pyridin-2-yl)prop-2-en-1-one

  • MDL number MFCD01315417
  • Cas Number : 18451-44-0
  • Molecular Weight : 239.274

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/72070

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3N-030_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart