KEY465195084 Key Organics / BIONET

(E)-N-[(4-methylphenyl)methyl]-2-[4-(propan-2-yl)phenyl]ethene-1-sulfonamide

  • MDL number MFCD01815107
  • Cas Number : 478032-19-8
  • Molecular Weight : 329.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/72074

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3N-034_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart