KEY465195087 Key Organics / BIONET

(2E)-3-[(4-bromophenyl)carbamoyl]prop-2-enoic acid

  • MDL number MFCD00027402
  • Cas Number : 843664-90-4
  • Molecular Weight : 270.082

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/72080

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3N-040_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart