KEY465195088 Key Organics / BIONET

(2E)-3-(2H-1,3-benzodioxol-5-yl)-1-(4-chloro-3-nitrophenyl)prop-2-en-1-one

  • MDL number MFCD01315420
  • Cas Number : 338415-88-6
  • Molecular Weight : 331.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/72082

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3N-042_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart