KEY465195089 Key Organics / BIONET

5-[2-(4-chloro-3-nitrobenzoyl)cyclopropyl]-2H-1,3-benzodioxole

  • MDL number MFCD01315530
  • Cas Number : 338415-90-0
  • Molecular Weight : 345.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/72084

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3N-044_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart