KEY465195090 Key Organics / BIONET

2-[2-(4-methoxyphenyl)cyclopropanecarbonyl]pyridine

  • MDL number MFCD01315531
  • Cas Number : 338415-92-2
  • Molecular Weight : 253.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/72087

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3N-047_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart