KEY465195094 Key Organics / BIONET

1-methyl-2-({[3-(trifluoromethyl)phenyl]methyl}sulfanyl)-1H-indole-3-carbaldehyde

  • MDL number MFCD01568346
  • Cas Number : 338416-41-4
  • Molecular Weight : 349.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/72116

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3N-319S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart