KEY465195103 Key Organics / BIONET

methyl 4-[(1E)-(hydroxyimino)methyl]-5-[(4-methylphenyl)sulfanyl]-1-phenyl-1H-pyrazole-3-carboxylate

  • MDL number MFCD01315842
  • Cas Number : 318238-01-6
  • Molecular Weight : 367.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/72191

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3N-554S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart