KEY465195104 Key Organics / BIONET

methyl 5-[(4-chlorophenyl)sulfanyl]-4-cyano-1-phenyl-1H-pyrazole-3-carboxylate

  • MDL number MFCD04125120
  • Cas Number : 956324-48-4
  • Molecular Weight : 369.82

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/72195

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3N-562S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart