KEY465195108 Key Organics / BIONET

methyl 5-(4-chlorophenyl)-1-phenyl-1H-pyrazole-3-carboxylate

  • MDL number MFCD01315848
  • Cas Number : 318238-06-1
  • Molecular Weight : 312.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/72222

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3N-609S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart