KEY465195111 Key Organics / BIONET

4-[4-methyl-2-(methylsulfanyl)-5H,6H,7H-pyrrolo[2,3-d]pyrimidin-7-yl]phenol

  • MDL number MFCD01316138
  • Cas Number : 338416-14-1
  • Molecular Weight : 273.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/72244

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3N-731_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart