KEY465195113 Key Organics / BIONET

N-tert-butyl-decahydroisoquinoline-3-carboxamide

  • MDL number MFCD01315994
  • Cas Number : 168899-60-3
  • Molecular Weight : 238.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name N-(tert-butyl)decahydro-3-isoquinolinecarboxamide
Inchi Code 1S/C14H26N2O/c1-14(2,3)16-13(17)12-8-10-6-4-5-7-11(10)9-15-12/h10-12,15H,4-9H2,1-3H3,(H,16,17)
Inchi Key UPZBXVBPICTBDP-UHFFFAOYSA-N

Description

Purity 90%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/72258

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/3N-745_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart