KEY465195116 Key Organics / BIONET

2,2-dimethyl-5-[(3-phenyl-1H-pyrazol-4-yl)methylidene]-1,3-dioxane-4,6-dione

  • MDL number MFCD01814822
  • Cas Number : 477711-33-4
  • Molecular Weight : 298.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/72270

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3P-009_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart