KEY465195118 Key Organics / BIONET

N'-[(1E)-[2-(benzenesulfonyl)-1,3-thiazol-5-yl]methylidene]furan-2-carbohydrazide

  • MDL number MFCD01814825
  • Cas Number : 478032-38-1
  • Molecular Weight : 361.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/72273

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3P-013_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart