KEY465195121 Key Organics / BIONET

methyl 4-[(E)-{[2-(benzenesulfonyl)-1,3-thiazol-5-yl]methylidene}amino]benzoate

  • MDL number MFCD01814830
  • Cas Number : 478032-42-7
  • Molecular Weight : 386.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/72279

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3P-022_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart