KEY465195127 Key Organics / BIONET

N-(4-ethoxyphenyl)-4-(thiophen-3-yl)-1,3-thiazol-2-amine

  • MDL number MFCD27924211
  • Cas Number : 1797206-90-6
  • Molecular Weight : 302.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 138 - 140
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/402882

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3P-054_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart