KEY465195128 Key Organics / BIONET

4-[(1E)-[2-(4-chlorophenyl)hydrazin-1-ylidene]methyl]-3-(4-fluorophenyl)-1H-pyrazole

  • MDL number MFCD01814841
  • Cas Number : 477711-38-9
  • Molecular Weight : 314.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/72302

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3P-062_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart