KEY465195130 Key Organics / BIONET

7-chloro-4-[(Z)-2-{[3-(4-fluorophenyl)-1H-pyrazol-4-yl]methylidene}hydrazin-1-yl]quinoline

  • MDL number MFCD01814843
  • Cas Number : 477711-39-0
  • Molecular Weight : 365.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/72305

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3P-066_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart