KEY465195132 Key Organics / BIONET

2,4-dichloro-1-[2-(chloromethyl)phenoxy]benzene

  • MDL number MFCD01871486
  • Cas Number : 478032-58-5
  • Molecular Weight : 287.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/72308

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3P-069_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart