KEY465195138 Key Organics / BIONET

N-(1-butyl-3-cyano-4,5-diphenyl-1H-pyrrol-2-yl)-2-methylpropanamide

  • MDL number MFCD01815198
  • Cas Number : 478032-68-7
  • Molecular Weight : 385.51

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/72330

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3P-307S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart