KEY465195140 Key Organics / BIONET

N-(1-benzyl-3-cyano-4,5-dimethyl-1H-pyrrol-2-yl)-4-fluorobenzamide

  • MDL number MFCD02082184
  • Cas Number : 478032-92-7
  • Molecular Weight : 347.393

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/72365

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3P-352S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart