KEY465195142 Key Organics / BIONET

N-(1-benzyl-3-cyano-4,5-dimethyl-1H-pyrrol-2-yl)thiophene-2-carboxamide

  • MDL number MFCD01815214
  • Cas Number : 478032-95-0
  • Molecular Weight : 335.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/72368

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3P-355S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart