KEY465195146 Key Organics / BIONET

N-(4-cyano-3-methyl-1,2-oxazol-5-yl)-4-fluorobenzamide

  • MDL number MFCD01815435
  • Cas Number : 478033-14-6
  • Molecular Weight : 245.213

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/72389

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3P-393S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart