KEY465195147 Key Organics / BIONET

N-(4-cyano-3-methyl-1,2-oxazol-5-yl)-3-methylbenzamide

  • MDL number MFCD04125132
  • Cas Number : 861212-51-3
  • Molecular Weight : 241.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/72390

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3P-394S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart