KEY465195154 Key Organics / BIONET

2-chloro-N-[2-(1,3-dimethyl-5-phenoxy-1H-pyrazol-4-yl)ethyl]acetamide

  • MDL number MFCD01815029
  • Cas Number : 477711-44-7
  • Molecular Weight : 307.78

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/72412

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3P-563S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart