KEY465195162 Key Organics / BIONET

2-[(E)-[(4-hydroxy-3-methoxyphenyl)methylidene]amino]-5-phenylfuran-3-carbonitrile

  • MDL number MFCD01815461
  • Cas Number : 478033-44-2
  • Molecular Weight : 318.332

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/72444

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3P-607S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart