KEY465195164 Key Organics / BIONET

N'-(4-methylbenzenesulfonyl)-4H,5H-naphtho[1,2-b]thiophene-2-carbohydrazide

  • MDL number MFCD01815469
  • Cas Number : 478033-50-0
  • Molecular Weight : 398.50

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/72452

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3P-643S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart