KEY465195166 Key Organics / BIONET

3-[2-(4-methylphenyl)-5-oxo-5,6-dihydro-4H-1,3,4-oxadiazin-4-yl]propanenitrile

  • MDL number MFCD04125144
  • Cas Number : 861212-61-5
  • Molecular Weight : 243.266

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/72454

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3P-645S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart