KEY465195173 Key Organics / BIONET

ethyl 2-[(2-acetyl-3-oxobut-1-en-1-yl)amino]acetate

  • MDL number MFCD01814847
  • Cas Number : 188782-93-6
  • Molecular Weight : 213.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/72481

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3P-710_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart