KEY465195176 Key Organics / BIONET

4-hydroxy-1-[2-(1H-indol-3-yl)ethyl]-3-[(2E)-3-phenylprop-2-en-1-yl]-1,2-dihydropyridin-2-one

  • MDL number MFCD02082510
  • Cas Number : 478033-74-8
  • Molecular Weight : 370.452

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/72488

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3P-720_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart