KEY465195205 Key Organics / BIONET

(1Z)-1-[4-(1H-1,2,3,4-tetrazol-5-yl)phenyl]-N-(4H-1,2,4-triazol-4-yl)methanimine

  • MDL number MFCD02186138
  • Cas Number : 478039-39-3
  • Molecular Weight : 240.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/72586

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3R-0121_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart