KEY465195217 Key Organics / BIONET

1-(4-{2-[methyl(pyridin-2-yl)amino]ethoxy}phenyl)ethan-1-ol

  • MDL number MFCD02661716
  • Cas Number : 439097-54-8
  • Molecular Weight : 272.348

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/72630

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3R-0189_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart