KEY465195219 Key Organics / BIONET

1-(4-bromophenyl)-2-[(1-phenyl-1H-imidazol-2-yl)sulfanyl]ethan-1-one

  • MDL number MFCD02186184
  • Cas Number : 478039-78-0
  • Molecular Weight : 373.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/72636

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3R-0197_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart