KEY465195223 Key Organics / BIONET

4-{3-[(2-chloro-6-fluorophenyl)methoxy]phenyl}pyrimidin-2-amine

  • MDL number MFCD02571896
  • Cas Number : 478039-82-6
  • Molecular Weight : 329.76

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/72642

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3R-0205_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart