KEY465195236 Key Organics / BIONET

2-[(2,4,6-trichlorophenyl)amino]-2,3,3a,4,7,7a-hexahydro-1H-isoindole-1,3-dione

  • MDL number MFCD01871202
  • Cas Number : 350680-03-4
  • Molecular Weight : 345.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/72692

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3R-0275_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart