KEY465195241 Key Organics / BIONET

4-[2-(4-chlorophenyl)acetyl]-N-[(pyridin-3-yl)methyl]-1H-pyrrole-2-carboxamide

  • MDL number MFCD02571904
  • Cas Number : 478040-33-4
  • Molecular Weight : 353.81

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/72706

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3R-0299_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart