KEY465195245 Key Organics / BIONET

4-benzoyl-N-[3-(1H-imidazol-1-yl)propyl]-1H-pyrrole-2-carboxamide

  • MDL number MFCD02186425
  • Cas Number : 478040-38-9
  • Molecular Weight : 322.368

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/72712

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3R-0307_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart