KEY465195264 Key Organics / BIONET

(3E)-4-(2,3-dihydro-1H-indol-1-yl)-1,1,1-trifluorobut-3-en-2-one

  • MDL number MFCD02186219
  • Cas Number : 428842-30-2
  • Molecular Weight : 241.213

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/72764

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3R-0674_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart