KEY465195266 Key Organics / BIONET

(3E)-4-[(1,3-benzothiazol-2-yl)amino]-1,1,1-trifluorobut-3-en-2-one

  • MDL number MFCD02186221
  • Cas Number : 478040-77-6
  • Molecular Weight : 272.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/72769

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3R-0686_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart