KEY465195268 Key Organics / BIONET

4-[(1,3-benzothiazol-2-yl)amino]-1,1,1-trifluorobutan-2-ol

  • MDL number MFCD04125150
  • Cas Number : 861212-67-1
  • Molecular Weight : 276.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/72772

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/3R-0690_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart