KEY465195270 Key Organics / BIONET

1-[1-(2-phenylethyl)-1H-1,3-benzodiazol-2-yl]ethan-1-one

  • MDL number MFCD02186509
  • Cas Number : 478040-86-7
  • Molecular Weight : 264.328

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/72780

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3R-0705_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart