KEY465195273 Key Organics / BIONET

(3Z)-4-(2,3-dihydro-1H-indol-1-yl)-1,1-difluorobut-3-en-2-one

  • MDL number MFCD02186227
  • Cas Number : 478040-92-5
  • Molecular Weight : 223.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/72787

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3R-0713_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart